missing translation for 'onlineSavingsMsg'
Learn More

Methyl dimethoxyacetate, 96%

CAS: 89-91-8 | C5H10O4 | 134.131 g/mol

$126.05 - $1895.93

Chemical Identifiers

CAS 89-91-8
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.131
MDL Number MFCD00008484
InChI Key NZTCVGHPDWAALP-UHFFFAOYSA-N
Synonym methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate
PubChem CID 66647
IUPAC Name methyl 2,2-dimethoxyacetate
SMILES COC(C(=O)OC)OC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1217109
View Documents
Thermo Scientific Chemicals
A1217109
10 g
Each for $126.05
Only null left
 
AAA1217118
View Documents
Thermo Scientific Chemicals
A1217118
50 g
Each for $497.82
Only null left
 
AAA1217130
View Documents
Thermo Scientific Chemicals
A1217130
250 g
Each for $1,895.93
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

89-91-8
134.131
NZTCVGHPDWAALP-UHFFFAOYSA-N
66647
COC(C(=O)OC)OC
C5H10O4
MFCD00008484
methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate
methyl 2,2-dimethoxyacetate

Specifications

89-91-8
160°C to 163°C
C5H10O4
MFCD00008484
1757582
methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate
COC(C(=O)OC)OC
134.131
134.13
Methyl dimethoxyacetate
1.098
63°C (145°F)
1.405
10 g
Moisture sensitive
NZTCVGHPDWAALP-UHFFFAOYSA-N
methyl 2,2-dimethoxyacetate
66647
≥97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 201-950-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only