Learn More
Methyl dimethoxyacetate, 96%
CAS: 89-91-8 | C5H10O4 | 134.131 g/mol
Supplier: Thermo Scientific Chemicals A1217118
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Methyl dimethoxyacetate | |
| 1.098 | |
| 63°C (145°F) | |
| 1.405 | |
| 50 g | |
| Moisture sensitive | |
| NZTCVGHPDWAALP-UHFFFAOYSA-N | |
| methyl 2,2-dimethoxyacetate | |
| 66647 | |
| ≥97% |
| 89-91-8 | |
| 160°C to 163°C | |
| C5H10O4 | |
| MFCD00008484 | |
| 1757582 | |
| methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate | |
| COC(C(=O)OC)OC | |
| 134.131 | |
| 134.13 |
Chemical Identifiers
| 89-91-8 | |
| 134.131 | |
| NZTCVGHPDWAALP-UHFFFAOYSA-N | |
| 66647 | |
| COC(C(=O)OC)OC |
| C5H10O4 | |
| MFCD00008484 | |
| methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate | |
| methyl 2,2-dimethoxyacetate |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 201-950-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only