missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Dihydrojasmonate (cis- and trans- mixture) 95.0+%, TCI America™
$95.06 - $95.06
Chemical Identifiers
| CAS | 24851-98-7 |
|---|---|
| Molecular Formula | C13H22O3 |
| Molecular Weight (g/mol) | 226.316 |
| MDL Number | MFCD00151188 |
| InChI Key | KVWWIYGFBYDJQC-UHFFFAOYSA-N |
| Synonym | 2-Amylcyclopentan-1-one-3-acetic Acid Methyl Ester, Dihydrojasmonic Acid Methyl Ester, Methyl 3-Oxo-2-pentyl-1-cyclopentaneacetate |
| PubChem CID | 102861 |
| IUPAC Name | methyl 2-(3-oxo-2-pentylcyclopentyl)acetate |
| SMILES | CCCCCC1C(CCC1=O)CC(=O)OC |
Chemical Identifiers
| 24851-98-7 | |
| 226.316 | |
| KVWWIYGFBYDJQC-UHFFFAOYSA-N | |
| 102861 | |
| CCCCCC1C(CCC1=O)CC(=O)OC |
| C13H22O3 | |
| MFCD00151188 | |
| 2-Amylcyclopentan-1-one-3-acetic Acid Methyl Ester, Dihydrojasmonic Acid Methyl Ester, Methyl 3-Oxo-2-pentyl-1-cyclopentaneacetate | |
| methyl 2-(3-oxo-2-pentylcyclopentyl)acetate |
Specifications
| 24851-98-7 | |
| 110°C | |
| MFCD00151188 | |
| 2-Amylcyclopentan-1-one-3-acetic Acid Methyl Ester, Dihydrojasmonic Acid Methyl Ester, Methyl 3-Oxo-2-pentyl-1-cyclopentaneacetate | |
| CCCCCC1C(CCC1=O)CC(=O)OC | |
| 226.316 | |
| 226.32 | |
| Liquid |
| Yellow | |
| C13H22O3 | |
| 25 mL | |
| KVWWIYGFBYDJQC-UHFFFAOYSA-N | |
| methyl 2-(3-oxo-2-pentylcyclopentyl)acetate | |
| 102861 | |
| ≥95.0% (GC) | |
| Methyl Dihydrojasmonate (cis- and trans- mixture) |
Safety and Handling
EINECSNumber : (3)-3309
RTECSNumber : GY2453800
TSCA : Yes