missing translation for 'onlineSavingsMsg'
Learn More

Methyl Difluoroacetate 97.0+%, TCI America™

Catalog No. p-7132973
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20215G 5 g
M202125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20215G Supplier TCI America Supplier No. M20215G
Only null left

Chemical Identifiers

CAS 433-53-4
Molecular Formula C3H4F2O2
Molecular Weight (g/mol) 110.06
MDL Number MFCD00039276
InChI Key CSSYKHYGURSRAZ-UHFFFAOYSA-N
Synonym methyl difluoroacetate, methyl2,2-difluoroacetate, difluoroacetic acid methyl ester, acetic acid, difluoro-, methyl ester, methyldifluoroacetate, methyl difluoroacetate #, acmc-209ju0, methyl 2,2-difluoroethanoate
PubChem CID 79012
IUPAC Name methyl 2,2-difluoroacetate
SMILES COC(=O)C(F)F

Specifications

Color Yellow
Boiling Point 86°C
Quantity 5 g
UN Number 2924
Formula Weight 110.06
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Difluoroacetate