missing translation for 'onlineSavingsMsg'
Learn More

Methyl Dichlorophosphite 95.0+%, TCI America™

Catalog No. M090510G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M090510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M090510G Supplier TCI America Supplier No. M090510G

Chemical Identifiers

CAS 3279-26-3
Molecular Formula CH3Cl2OP
Molecular Weight (g/mol) 132.908
MDL Number MFCD00000524
InChI Key HCSDJECSMANTCX-UHFFFAOYSA-N
Synonym methyl dichlorophosphite, methyl phosphorodichloridite, methyl dichlorophosphinite, dichloro methyl phosphite, phosphorodichloridous acid, methyl ester, methyldichlorophosphite, methoxydichlorophosphine, ch3opcl2, dichloro methoxy phosphane, dichloro methoxy phosphine
PubChem CID 76768
IUPAC Name dichloro(methoxy)phosphane
SMILES COP(Cl)Cl

Specifications

Color Colorless
Boiling Point 92°C
Quantity 10 g
UN Number 2920
Formula Weight 132.91
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Methyl Dichlorophosphite [Phosphitylating Agent]
TSCA No
Recommended Storage Refrigerator