accessibility menu, dialog, popup

UserName

H3COPClCl

CAS RN 3279-26-3

IUPAC Name: methyl dichlorophosphinite
Synonyms: methyl dichlorophosphinite
Molecular Weight (g/mol): 132.91
Molecular Formula: CH3Cl2OP
InChi Key: HCSDJECSMANTCX-UHFFFAOYSA-N
SMILES: COP(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results