missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl β-D-thiogalactoside, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 155-30-6 |
|---|---|
| Molecular Formula | C7H14O5S |
| Molecular Weight (g/mol) | 210.25 |
| InChI Key | LZFNFLTVAMOOPJ-PZRMXXKTSA-N |
| Synonym | methyl-beta-d-thiogalactoside, methyl-1-thio-beta-d-galactopyranoside, methyl 1-thio-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methylsulfanyl oxane-3,4,5-triol, b-d-thiomethylgalactoside, methyl b-d-thiogalactoside, galactose-derived compound 19, methyl beta-d-thio-galactopyranoside, methyl 1-thio-??-d-galactopyranoside, 1-s-methyl-1-thio-beta-d-galactopyranose |
| PubChem CID | 10036019 |
| IUPAC Name | (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methylsulfanyloxane-3,4,5-triol |
| SMILES | CSC1C(C(C(C(O1)CO)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC459452500
|
N/A | 250mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC459450010
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 155-30-6 | |
| 210.25 | |
| methyl-beta-d-thiogalactoside, methyl-1-thio-beta-d-galactopyranoside, methyl 1-thio-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methylsulfanyl oxane-3,4,5-triol, b-d-thiomethylgalactoside, methyl b-d-thiogalactoside, galactose-derived compound 19, methyl beta-d-thio-galactopyranoside, methyl 1-thio-??-d-galactopyranoside, 1-s-methyl-1-thio-beta-d-galactopyranose | |
| (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methylsulfanyloxane-3,4,5-triol |
| C7H14O5S | |
| LZFNFLTVAMOOPJ-PZRMXXKTSA-N | |
| 10036019 | |
| CSC1C(C(C(C(O1)CO)O)O)O |
Specifications
| 155-30-6 | |
| 97.5% min. (HPLC) | |
| methyl-beta-d-thiogalactoside, methyl-1-thio-beta-d-galactopyranoside, methyl 1-thio-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methylsulfanyl oxane-3,4,5-triol, b-d-thiomethylgalactoside, methyl b-d-thiogalactoside, galactose-derived compound 19, methyl beta-d-thio-galactopyranoside, methyl 1-thio-??-d-galactopyranoside, 1-s-methyl-1-thio-beta-d-galactopyranose | |
| LZFNFLTVAMOOPJ-PZRMXXKTSA-N | |
| (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methylsulfanyloxane-3,4,5-triol | |
| 10036019 | |
| 98% |
| Authentic | |
| C7H14O5S | |
| Solubility in water: soluble. | |
| CSC1C(C(C(C(O1)CO)O)O)O | |
| 210.25 | |
| 210.25 | |
| Methyl β-D-thiogalactoside |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Causes damage to organs.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
GHS Signal Word: Danger
EINECSNumber : 205-842-4
RUO – Research Use Only