accessibility menu, dialog, popup

UserName

CAS RN 155-30-6

H3CS(S)OOH(R)OHHO(R)HO

CAS RN 155-30-6

IUPAC Name: (3R,5R,6S)-2-(hydroxymethyl)-6-(methylsulfanyl)oxane-3,4,5-triol
Synonyms: (3R,5R,6S)-2-(hydroxymethyl)-6-(methylsulfanyl)oxane-3,4,5-triol
Molecular Weight (g/mol): 210.24
Molecular Formula: C7H14O5S
InChi Key: LZFNFLTVAMOOPJ-XMAYFYEJSA-N
SMILES: CS[C@@H]1OC(CO)[C@H](O)C(O)[C@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)