missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl α-D-mannopyranoside, ≥99%, Fisher BioReagents™
$283.84 - $4916.37
Chemical Identifiers
| CAS | 617-04-9 |
|---|---|
| Molecular Formula | C7H14O6 |
| Molecular Weight (g/mol) | 194.18 |
| MDL Number | MFCD00063262 |
| InChI Key | HOVAGTYPODGVJG-UHFFFAOYNA-N |
| Synonym | methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside |
| PubChem CID | 101798 |
| ChEBI | CHEBI:43943 |
| SMILES | COC1OC(CO)C(O)C(O)C1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
BP253010
|
Fisher BioReagents
BP253010 |
10 g | Amber Glass |
Each for $283.84
Case of 8 Each for $2,270.72
|
|
||||
|
BP2530100
|
Fisher BioReagents
BP2530100 |
100 g | Amber Glass |
Each for $635.05
Case of 3 Each for $1,905.15
|
|
||||
|
BP2530500
|
Fisher BioReagents
BP2530500 |
500 g | Amber Glass |
Each for $4,916.37
Case of 3 Each for $14,749.11
|
|
||||
Description
Methyl α-D-mannopyranoside, ≥99%
Methyl α-D-Mannopyranoside is used in biochemical research applications.Chemical Identifiers
| 617-04-9 | |
| 194.18 | |
| HOVAGTYPODGVJG-UHFFFAOYNA-N | |
| 101798 | |
| COC1OC(CO)C(O)C(O)C1O |
| C7H14O6 | |
| MFCD00063262 | |
| methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside | |
| CHEBI:43943 |
Specifications
| 617-04-9 | |
| White | |
| ≥99% | |
| C7H14O6 | |
| 10 g | |
| methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside | |
| COC1OC(CO)C(O)C(O)C1O | |
| 194.18 | |
| CHEBI:43943 | |
| ≥99% | |
| Methyl α-D-Mannopyranoside |
| 193°C to 196°C | |
| < 0.5% | |
| Amber Glass | |
| MFCD00063262 | |
| 76 to 85° (+ or -) | |
| HOVAGTYPODGVJG-UHFFFAOYNA-N | |
| (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol | |
| 101798 | |
| 194.2 | |
| Powder or Solid |
Safety and Handling
Recommended Storage : RT