missing translation for 'onlineSavingsMsg'
Learn More

Methyl α-D-mannopyranoside, ≥99%, Fisher BioReagents™

Supplier:  Fisher BioReagents BP2530100

 View more versions of this product

Catalog No. BP2530100



Only null left

Description

Description

Methyl α-D-mannopyranoside, ≥99%

  • Methyl α-D-mannopyranoside is a lectin inhibitory sugar
  • Competitive inhibitor of the binding of mannose by E. coli
  •  Intended for research use only
Specifications

Specifications

Methyl α-D-Mannopyranoside
193°C to 196°C
< 0.5%
Amber Glass
MFCD00063262
76 to 85° (+ or -)
HOVAGTYPODGVJG-UHFFFAOYNA-N
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
101798
194.2
Powder or Solid
617-04-9
White
≥99%
C7H14O6
100 g
methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
COC1OC(CO)C(O)C(O)C1O
194.18
CHEBI:43943
≥99%

Chemical Identifiers

617-04-9
194.18
HOVAGTYPODGVJG-UHFFFAOYNA-N
101798
COC1OC(CO)C(O)C(O)C1O
C7H14O6
MFCD00063262
methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
CHEBI:43943
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS Documents

Documents

Promotions

Promotions