missing translation for 'onlineSavingsMsg'
Learn More

Methyl α-D-Mannopyranoside, 99.3%, MP Biomedicals

Supplier:  MP Biomedicals Inc 0215393725

 View more versions of this product

Catalog No. ICN15393725


Only null left

Description

Description

Specifications

Specifications

Methyl α-D-Mannopyranoside
White
C7H14O6
25 g
HOVAGTYPODGVJG-UHFFFAOYNA-N
2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
101798
194.2
617-04-9
∽ 99%
MFCD00063262
methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
COC1OC(CO)C(O)C(O)C1O
194.18
CHEBI:43943
Crystalline Powder

Chemical Identifiers

617-04-9
194.18
HOVAGTYPODGVJG-UHFFFAOYNA-N
101798
2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
C7H14O6
MFCD00063262
methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
CHEBI:43943
COC1OC(CO)C(O)C(O)C1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at room temperature.

SDS
Documents

Documents

Promotions

Promotions