missing translation for 'onlineSavingsMsg'
Learn More

Methyl D-(+)-Lactate 98.0+%, TCI America™

Supplier:  TCI America L013625G

 View more versions of this product

Catalog No. L013625G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methyl D-(+)-Lactate
Colorless
C4H8O3
25 g
methyl r-+-lactate, methyl d-lactate, r-methyl 2-hydroxypropanoate, methyl r-lactate, +-methyl d-lactate, methyl 2r-2-hydroxypropanoate, d-lactic acid methyl ester, methyl lactate, +, unii-45mz1t3tbv, r-methyl lactate
COC(=O)C(C)O
104.11
CHEBI:74611
≥98.0% (GC)
17392-83-5
33°C
MFCD00004517
3272
LPEKGGXMPWTOCB-UHFFFAOYNA-N
methyl 2-hydroxypropanoate
637514
104.11
Liquid

Chemical Identifiers

17392-83-5
104.11
LPEKGGXMPWTOCB-UHFFFAOYNA-N
637514
methyl 2-hydroxypropanoate
C4H8O3
MFCD00004517
methyl r-+-lactate, methyl d-lactate, r-methyl 2-hydroxypropanoate, methyl r-lactate, +-methyl d-lactate, methyl 2r-2-hydroxypropanoate, d-lactic acid methyl ester, methyl lactate, +, unii-45mz1t3tbv, r-methyl lactate
CHEBI:74611
COC(=O)C(C)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1370

TSCA : No

SDS
Documents

Documents

Promotions

Promotions