missing translation for 'onlineSavingsMsg'
Learn More

Methyl D-(+)-Lactate 98.0+%, TCI America™

Catalog No. p-7131834
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01365G 5 g
L013625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L01365G Supplier TCI America Supplier No. L01365G
Only null left

Chemical Identifiers

CAS 17392-83-5
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00004517
InChI Key LPEKGGXMPWTOCB-UHFFFAOYNA-N
Synonym methyl r-+-lactate, methyl d-lactate, r-methyl 2-hydroxypropanoate, methyl r-lactate, +-methyl d-lactate, methyl 2r-2-hydroxypropanoate, d-lactic acid methyl ester, methyl lactate, +, unii-45mz1t3tbv, r-methyl lactate
PubChem CID 637514
ChEBI CHEBI:74611
IUPAC Name methyl 2-hydroxypropanoate
SMILES COC(=O)C(C)O

Specifications

Color Colorless
Boiling Point 33°C
Quantity 5 g
UN Number 3272
Formula Weight 104.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl D-(+)-Lactate
EINECSNumber (2)-1370
TSCA No