missing translation for 'onlineSavingsMsg'
Learn More

Methyl cyclopropanecarboxylate, 98%

Catalog No. AAB2085322
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2085322 100 g
AAB2085314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2085322 Supplier Thermo Scientific Chemicals Supplier No. B2085322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2868-37-3 | C5H8O2 | 100.117 g/mol

It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. Methyl cyclopropane carboxylate is used as a reagent in the synthesis of N’-thiazole and N-pyrazole based inhibitors of MAP kinase p38α.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. Methyl cyclopropane carboxylate is used as a reagent in the synthesis of N’-thiazole and N-pyrazole based inhibitors of MAP kinase p38α.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents and ignition sources. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges.

Chemical Identifiers

CAS 2868-37-3
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00001280
InChI Key PKAHQJNJPDVTDP-UHFFFAOYSA-N
Synonym methyl cyclopropane carboxylate, cyclopropanecarboxylic acid, methyl ester, methylcyclopropanecarboxylate, cyclopropanecarboxylic acid methyl ester, methyl cyclopropylcarboxylate, unii-5uq53f585u, mcpc, pubchem11835, ksc490a7l, 1-methylcyclopropanecarboxylate
PubChem CID 76122
IUPAC Name methyl cyclopropanecarboxylate
SMILES COC(=O)C1CC1

Specifications

Density 0.983
Boiling Point 118°C to 119°C
Flash Point 22°C (71°F)
Refractive Index 1.418
Quantity 100 g
UN Number UN3272
Beilstein 2039388
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 100.12
Percent Purity 98%
Chemical Name or Material Methyl cyclopropanecarboxylate
Show More Show Less
Hazard Category H225-H302-H311-H319
Hazard Statement GHS H Statement
H225-H331-H302-H319
Highly flammable liquid and vapour.
Toxic if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P243-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P362-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 220-690-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only