accessibility menu, dialog, popup

UserName

CAS RN 2868-37-3

IUPAC Name: methyl cyclopropanecarboxylate
Synonyms: cyclopropanecarboxylic acid, methyl ester
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: PKAHQJNJPDVTDP-UHFFFAOYSA-N
SMILES: COC(=O)C1CC1
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)