missing translation for 'onlineSavingsMsg'
Learn More

Methyl cyclopentanecarboxylate, 97%

Catalog No. AAL0074114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0074114 25 g
AAL0074106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0074114 Supplier Thermo Scientific Chemicals Supplier No. L0074114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4630-80-2 | C7H12O2 | 128.17 g/mol

Methyl cyclopentanecarboxylate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl cyclopentanecarboxylate

Solubility
Difficult to mix.

Notes
Protect from heat. Store away from oxidizing agents.

Chemical Identifiers

CAS 4630-80-2
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00019290
InChI Key IIHIJFJSXPDTNO-UHFFFAOYSA-N
Synonym cyclopentanecarboxylic acid, methyl ester, cyclopentanecarboxylic acid methyl ester, methyl cyclopentanoate, acmc-1am5n, methylcyclopentanecarboxylate, methylcyclopentane carboxylate, methyl cyclopentane carboxylate, methyl-1-cyclopentanecarboxylate, cyclopentanoic acid methyl ester, cyclopentanecarboxylic acid methyl
PubChem CID 78365
IUPAC Name methyl cyclopentanecarboxylate
SMILES COC(=O)C1CCCC1

Specifications

Boiling Point 81°C to 82°C (5 mmHg)
Refractive Index 1.436
Quantity 25 g
Beilstein 2040762
Solubility Information Difficult to mix.
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material Methyl cyclopentanecarboxylate
Hazard Category H226-H302
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P330-P374-P380-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 225-049-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only