accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 4630-80-2

IUPAC Name: methyl cyclopentanecarboxylate
Synonyms: methyl cyclopentanecarboxylate
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: IIHIJFJSXPDTNO-UHFFFAOYSA-N
SMILES: COC(=O)C1CCCC1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)