missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Cyclohexanecarboxylate 98.0+%, TCI America™
Supplier: TCI America C0471100ML
Specifications
| Methyl Cyclohexanecarboxylate | |
| Colorless | |
| C8H14O2 | |
| 100 mL | |
| cyclohexanecarboxylic acid, methyl ester, methyl cyclohexanoate, methyl cyclohexylcarboxylate, cyclohexanecarboxylic acid methyl ester, hexahydrobenzoic acid methyl ester, methyl cyclohexylformate, methyl hexahydrobenzoate, methyl cyclohexane carboxylate, fema no. 3568, methyl ester of cyclohexanecarboxylic acid | |
| COC(=O)C1CCCCC1 | |
| 142.20 | |
| 142.20 | |
| Liquid |
| 4630-82-4 | |
| 183°C | |
| MFCD00001458 | |
| 3272 | |
| ZQWPRMPSCMSAJU-UHFFFAOYSA-N | |
| methyl cyclohexanecarboxylate | |
| 20748 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 4630-82-4 | |
| 142.20 | |
| ZQWPRMPSCMSAJU-UHFFFAOYSA-N | |
| 20748 | |
| COC(=O)C1CCCCC1 |
| C8H14O2 | |
| MFCD00001458 | |
| cyclohexanecarboxylic acid, methyl ester, methyl cyclohexanoate, methyl cyclohexylcarboxylate, cyclohexanecarboxylic acid methyl ester, hexahydrobenzoic acid methyl ester, methyl cyclohexylformate, methyl hexahydrobenzoate, methyl cyclohexane carboxylate, fema no. 3568, methyl ester of cyclohexanecarboxylic acid | |
| methyl cyclohexanecarboxylate |
Safety and Handling
RTECSNumber : GU8599000
TSCA : No