missing translation for 'onlineSavingsMsg'
Learn More

Methyl Cyclohexanecarboxylate 98.0+%, TCI America™

Catalog No. p-7124446
Change view
Click to view available options
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
C047125ML 25 mL
C0471100ML 100 mL
C0471500ML 500 mL
3 options

Catalog No. C047125ML

Supplier: TCI America C047125ML

Only null left

Chemical Identifiers

CAS 4630-82-4
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00001458
InChI Key ZQWPRMPSCMSAJU-UHFFFAOYSA-N
Synonym cyclohexanecarboxylic acid, methyl ester, methyl cyclohexanoate, methyl cyclohexylcarboxylate, cyclohexanecarboxylic acid methyl ester, hexahydrobenzoic acid methyl ester, methyl cyclohexylformate, methyl hexahydrobenzoate, methyl cyclohexane carboxylate, fema no. 3568, methyl ester of cyclohexanecarboxylic acid
PubChem CID 20748
IUPAC Name methyl cyclohexanecarboxylate
SMILES COC(=O)C1CCCCC1

Specifications

Color Colorless
Boiling Point 183°C
Quantity 25 mL
UN Number 3272
Formula Weight 142.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Cyclohexanecarboxylate
RTECSNumber GU8599000
TSCA No