missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Cyclobutanecarboxylate 98.0+%, TCI America™
Supplier: TCI America M288425G
Specifications
| Methyl Cyclobutanecarboxylate | |
| Colorless | |
| C6H10O2 | |
| 25 g | |
| cyclobutanecarboxylic acid methyl ester, cyclobutanecarboxylic acid, methyl ester, pubchem23839, methyl cyclobutane carboxylate, cyclobutanecarboxylicacid,methylester, cyclobutanecarboxylicacid, methyl ester, cyclobutane carboxylic acid methyl ester | |
| COC(=O)C1CCC1 | |
| 114.14 | |
| 114.14 | |
| Liquid |
| 765-85-5 | |
| 135°C | |
| MFCD00095142 | |
| 3272 | |
| CBTGNLZUIZHUHY-UHFFFAOYSA-N | |
| methyl cyclobutanecarboxylate | |
| 136594 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 765-85-5 | |
| 114.14 | |
| CBTGNLZUIZHUHY-UHFFFAOYSA-N | |
| 136594 | |
| COC(=O)C1CCC1 |
| C6H10O2 | |
| MFCD00095142 | |
| cyclobutanecarboxylic acid methyl ester, cyclobutanecarboxylic acid, methyl ester, pubchem23839, methyl cyclobutane carboxylate, cyclobutanecarboxylicacid,methylester, cyclobutanecarboxylicacid, methyl ester, cyclobutane carboxylic acid methyl ester | |
| methyl cyclobutanecarboxylate |
Safety and Handling
TSCA : No