missing translation for 'onlineSavingsMsg'
Learn More

Methyl Cyclobutanecarboxylate 98.0+%, TCI America™

Catalog No. p-7133489
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
M28845G 5 g
M288425G 25 g
2 options

Catalog No. M28845G

Supplier: TCI America M28845G

Only null left

Chemical Identifiers

CAS 765-85-5
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00095142
InChI Key CBTGNLZUIZHUHY-UHFFFAOYSA-N
Synonym cyclobutanecarboxylic acid methyl ester, cyclobutanecarboxylic acid, methyl ester, pubchem23839, methyl cyclobutane carboxylate, cyclobutanecarboxylicacid,methylester, cyclobutanecarboxylicacid, methyl ester, cyclobutane carboxylic acid methyl ester
PubChem CID 136594
IUPAC Name methyl cyclobutanecarboxylate
SMILES COC(=O)C1CCC1

Specifications

Color Colorless
Boiling Point 135°C
Quantity 5 g
UN Number 3272
Formula Weight 114.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Cyclobutanecarboxylate