missing translation for 'onlineSavingsMsg'
Learn More

Methyl Cyanoacetate 99.0+%, TCI America™

Supplier:  TCI America C0442500ML

 View more versions of this product

Catalog No. C0442500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methyl Cyanoacetate
Colorless
C4H5NO2
500 mL
ANGDWNBGPBMQHW-UHFFFAOYSA-N
methyl 2-cyanoacetate
7747
99.09
Liquid
105-34-0
205°C
MFCD00001939
methyl cyanoacetate, cyanoacetic acid methyl ester, methyl cyanoethanoate, acetic acid, cyano-, methyl ester, usaf kf-22, methylester kyseliny kyanoctove, methylcyanoacetate, malonic methyl ester nitrile, acetic acid, 2-cyano-, methyl ester, methyl cyano acetate
COC(=O)CC#N
99.09
CHEBI:51928
≥99.0% (GC)

Chemical Identifiers

105-34-0
99.09
ANGDWNBGPBMQHW-UHFFFAOYSA-N
7747
methyl 2-cyanoacetate
C4H5NO2
MFCD00001939
methyl cyanoacetate, cyanoacetic acid methyl ester, methyl cyanoethanoate, acetic acid, cyano-, methyl ester, usaf kf-22, methylester kyseliny kyanoctove, methylcyanoacetate, malonic methyl ester nitrile, acetic acid, 2-cyano-, methyl ester, methyl cyano acetate
CHEBI:51928
COC(=O)CC#N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1549

RTECSNumber : AG4375000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions