missing translation for 'onlineSavingsMsg'
Learn More

Methyl Cyanoacetate 99.0+%, TCI America™

Catalog No. p-7124423
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C044225ML 25 mL
C0442500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C044225ML Supplier TCI America Supplier No. C044225ML
Only null left

Chemical Identifiers

CAS 105-34-0
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.09
MDL Number MFCD00001939
InChI Key ANGDWNBGPBMQHW-UHFFFAOYSA-N
Synonym methyl cyanoacetate, cyanoacetic acid methyl ester, methyl cyanoethanoate, acetic acid, cyano-, methyl ester, usaf kf-22, methylester kyseliny kyanoctove, methylcyanoacetate, malonic methyl ester nitrile, acetic acid, 2-cyano-, methyl ester, methyl cyano acetate
PubChem CID 7747
ChEBI CHEBI:51928
IUPAC Name methyl 2-cyanoacetate
SMILES COC(=O)CC#N

Specifications

Color Colorless
Boiling Point 205°C
Quantity 25 mL
Formula Weight 99.09
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Cyanoacetate
EINECSNumber (2)-1549
RTECSNumber AG4375000
TSCA Yes