missing translation for 'onlineSavingsMsg'
Learn More

Methyl Crotonate 98.0+%, TCI America™

Catalog No. p-7124410
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Catalog No. Quantity
C041925ML 25 mL
C0419100ML 100 mL
C0419500ML 500 mL
3 options

Catalog No. C041925ML

Supplier: TCI America C041925ML

Only null left

Chemical Identifiers

CAS 623-43-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00009287
InChI Key MCVVUJPXSBQTRZ-ONEGZZNKSA-N
Synonym methyl crotonate, e-methyl but-2-enoate, trans-methyl crotonate, methyl e-crotonate, methyl 2-butenoate, methyl trans-crotonate, 2-butenoic acid, methyl ester, 2e, trans-2-butenoic acid methyl ester, methyl trans-2-butenoate, e-crotonic acid methyl ester
PubChem CID 638132
IUPAC Name methyl (2E)-but-2-enoate
SMILES COC(=O)\C=C\C

Specifications

Color Colorless
Boiling Point 120°C
Quantity 25 mL
UN Number 3272
Formula Weight 100.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Crotonate
EINECSNumber (2)-0966
RTECSNumber GQ5710000
TSCA Yes