missing translation for 'onlineSavingsMsg'
Learn More

Methyl cis-15-tetracosenoate, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013792
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013792 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013792 Supplier Merck Emd Millipore Supplier No. 17265100MG
Only null left

Methyl cis-15-tetracosenoate is a fatty acid methyl ester.

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 2733-88-2
Shipping Condition Ambient
Physical Form Neat
Linear Formula C25H48O2
MDL Number MFCD00056324
Synonym Nervonic acid methyl ester; Selacholeic acid methyl ester
Type Analytical Standard
Quantity 100 mg
Formula Weight 380.65
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less