missing translation for 'onlineSavingsMsg'
Learn More

Methyl cholate, 97%

Catalog No. AAL0512914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0512914 25 g
AAL0512906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0512914 Supplier Thermo Scientific Chemicals Supplier No. L0512914

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1448-36-8 | C25H42O5 | 422.606 g/mol

Methyl cholate, is used as a bile acid derivative as anti-inflammatory and antitumor agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl cholate, is used as a bile acid derivative as anti-inflammatory and antitumor agent.

Solubility
Insoluble in water. Soluble in medium polarity organic solvents.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 1448-36-8
Molecular Formula C25H42O5
Molecular Weight (g/mol) 422.606
MDL Number MFCD00064934
InChI Key DLYVTEULDNMQAR-BSEHQKOQSA-N
Synonym Cholic acid methyl ester
PubChem CID 133124937
IUPAC Name methyl (4S)-4-[(3R,7S,8S,9R,10R,12R,13S,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
SMILES CC(CCC(=O)OC)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C

Specifications

Melting Point 155°C to 156°C
Quantity 25 g
Beilstein 2226989
Merck Index 14,2203
Solubility Information Insoluble in water. Soluble in medium polarity organic solvents.
Formula Weight 422.61
Percent Purity ≥98%
Chemical Name or Material Methyl cholate
EINECSNumber 215-903-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only