missing translation for 'onlineSavingsMsg'
Learn More

Methyl Chloroglyoxylate 98.0+%, TCI America™

Catalog No. p-7125062
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C156125G 25 g
C1561250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C156125G Supplier TCI America Supplier No. C156125G
Only null left

Chemical Identifiers

CAS 5781-53-3
Molecular Formula C3H3ClO3
Molecular Weight (g/mol) 122.504
MDL Number MFCD00000705
InChI Key ZXUQEPZWVQIOJE-UHFFFAOYSA-N
Synonym methyl oxalyl chloride, methyl chloroglyoxylate, methyl chlorooxoacetate, chloroglyoxylic acid methyl ester, acetic acid, chlorooxo-, methyl ester, methyl chlorocarbonyl formate, methyloxalylchloride, methyl chloro oxo acetate, methylchlorooxoacetate, methyl 2-chloro-2-oxo-acetate
PubChem CID 79846
IUPAC Name methyl 2-chloro-2-oxoacetate
SMILES COC(=O)C(=O)Cl

Specifications

Color Yellow
Boiling Point 125°C
Quantity 25 g
UN Number 2920
Formula Weight 122.50
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Methyl Chloroglyoxylate