missing translation for 'onlineSavingsMsg'
Learn More

Methyl chlorodifluoroacetate, 98%

Catalog No. p-7024268
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
250 g
Catalog No. Quantity
AAA1862309 10 g
AAA1862330 250 g
2 options

Catalog No. AAA1862309

Supplier: Thermo Scientific Chemicals A1862309

Only null left

CAS: 1514-87-0 | C3H3ClF2O2 | 144.502 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1514-87-0
Molecular Formula C3H3ClF2O2
Molecular Weight (g/mol) 144.502
MDL Number MFCD00000775
InChI Key AWUPLMYXZJKHEG-UHFFFAOYSA-N
Synonym methyl chlorodifluoroacetate, chlorodifluoroacetic acid methyl ester, acetic acid, chlorodifluoro-, methyl ester, methyl difluorochloroacetic acid, methyl chloro difluoro acetate, acetic acid, 2-chloro-2,2-difluoro-, methyl ester, methyl-2-chloro-2,2-difluoroacetate, chloro-difluoro-acetic acid methyl ester, clcf2coome, clcf2co2me
PubChem CID 73935
IUPAC Name methyl 2-chloro-2,2-difluoroacetate
SMILES COC(=O)C(F)(F)Cl

Specifications

Density 1.371
Boiling Point 79°C to 81°C
Flash Point 19°C (66°F)
Refractive Index 1.35
Quantity 10 g
UN Number UN2924
Beilstein 1756072
Sensitivity Moisture sensitive
Formula Weight 144.5
Percent Purity 98%
Chemical Name or Material Methyl chlorodifluoroacetate
Show More Show Less
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 216-154-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only