missing translation for 'onlineSavingsMsg'
Learn More

Methyl Chloride (ca. 5.7% in Tetrahydrofuran, ca. 1mol/L), TCI America™

Supplier:  TCI America M2813100ML

Catalog No. M2813100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methyl Chloride (ca. 5.7% in Tetrahydrofuran, ca. 1mol/L)
Colorless
MFCD00000872
1993
NEHMKBQYUWJMIP-UHFFFAOYSA-N
chloromethane
6327
50.49
74-87-3
CH3Cl
100 mL
methyl chloride, methane, chloro, monochloromethane, methylchloride, artic, methylchlorid, clorometano, chloor-methaan, metylu chlorek, chlor-methan
CCl
50.485
CHEBI:36014
Liquid

Chemical Identifiers

74-87-3
50.485
NEHMKBQYUWJMIP-UHFFFAOYSA-N
6327
chloromethane
CH3Cl
MFCD00000872
methyl chloride, methane, chloro, monochloromethane, methylchloride, artic, methylchlorid, clorometano, chloor-methaan, metylu chlorek, chlor-methan
CHEBI:36014
CCl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0035&(5)-0053

RTECSNumber : PA6300000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions