accessibility menu, dialog, popup

UserName

CH3Cl

CAS RN 74-87-3

IUPAC Name: chloromethane
Synonyms: methyl chloride caswell no. 557 artic clorometano MeCl methylchlorid chloor-methaan C-H3-cl cloruro di metile rcra waste number U045
Molecular Weight (g/mol): 50.49
Molecular Formula: CH3Cl
InChi Key: NEHMKBQYUWJMIP-UHFFFAOYSA-N
SMILES: CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results