missing translation for 'onlineSavingsMsg'
Learn More

Methyl cellulose, viscosity 400 cP (2% solution in water)

CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol

Supplier:  Thermo Scientific Chemicals 428430010

Quantity 1 kg
Packaging Glass bottle

 View more versions of this product

Catalog No. AC428430010


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9004-67-5
454.513
YLGXILFCIXHCMC-JHGZEJCSSA-N
(5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
C20H38O11
MFCD00081763
51063134
COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifications

Methyl cellulose
9004-67-5
C20H38O11
MFCD00081763
Solubility in water: soluble in cold water. Other solubilities: soluble in glacial acetic acid,insoluble in acetone,alcohol,ether,chloroform,and toluene
COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
454.513
Viscosity 400 cP (2% solution in water)
Glass bottle
1 kg
15,6111
YLGXILFCIXHCMC-JHGZEJCSSA-N
(5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
51063134
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions