missing translation for 'onlineSavingsMsg'
Learn More

Methyl cellulose, MP Biomedicals™

Catalog No. p-3572973
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15549690 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15549690 Supplier MP Biomedicals Inc Supplier No. 0215549690

Methyl cellulose , White Powder

  • It is used as a substitute for water-soluble gums,to render paper grease proof in adhesives,as thickening agent in cosmetics,as protective colloid in emulsions,as binder and stabilizer in foods,as bulk producer in the formulation of dietetic foods,Pharmaceutic aid (suspending agent),Laxative.
  • It is a chemical compound derived from cellulose. It is a hydrophilic white powder in pure form and dissolves in cold (but not in hot) water, forming a clear viscous solution or gel.
  • It is used as a thickener and emulsifier in various food and cosmetic products. It is not digestible, not toxic, and not an allergen.

Chemical Identifiers

CAS 9004-67-5
Molecular Formula C20H38O11
MDL Number MFCD00081763
Synonym Cellulose methyl ether, Methocel MC, Cellothyl, Syncelose
PubChem CID 51063134
IUPAC Name (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifications

Color White
pH 6.7
Loss on Drying ≤5.0%
Quantity 500 g
Solubility Information Soluble in cold water. Insoluble in hot water.
InChI Key YLGXILFCIXHCMC-JHGZEJCSSA-N
Molecular Weight (g/mol) 454.513
Physical Form Powder
Chemical Name or Material Methyl Cellulose, Viscosity of 2% aqueous solution at 25°C: 4, 000cps
RTECSNumber FJ5959000
storageNote1 Store at Room Temperature (15-30°C).
Recommended Storage Room Temperature