missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl cellulose, MP Biomedicals™
Description
- It is used as a substitute for water-soluble gums,to render paper grease proof in adhesives,as thickening agent in cosmetics,as protective colloid in emulsions,as binder and stabilizer in foods,as bulk producer in the formulation of dietetic foods,Pharmaceutic aid (suspending agent),Laxative.
- It is a chemical compound derived from cellulose. It is a hydrophilic white powder in pure form and dissolves in cold (but not in hot) water, forming a clear viscous solution or gel.
- It is used as a thickener and emulsifier in various food and cosmetic products. It is not digestible, not toxic, and not an allergen.
Specifications
Specifications
| Color | White |
| pH | 6.7 |
| Loss on Drying | ≤5.0% |
| Quantity | 500 g |
| Solubility Information | Soluble in cold water. Insoluble in hot water. |
| InChI Key | YLGXILFCIXHCMC-JHGZEJCSSA-N |
| Molecular Weight (g/mol) | 454.513 |
| Grade | Cell Culture |
| Physical Form | Powder |
| Chemical Name or Material | Methyl Cellulose, Viscosity of 2% aqueous solution at 25°C: 4, 000cps |
Safety and Handling
| RTECSNumber | FJ5959000 |
| storageNote1 | Store at Room Temperature (15-30°C). |
| Recommended Storage | Room Temperature |