missing translation for 'onlineSavingsMsg'
Learn More

Methyl Cellulose, viscosity of 2% aqueous solution (25°C) 15cps, MP Biomedicals

$863.93 - $863.93

Chemical Identifiers

CAS 9004-67-5
Molecular Formula C20H38O11
Molecular Weight (g/mol) 454.513
InChI Key YLGXILFCIXHCMC-JHGZEJCSSA-N
Synonym Cellulose methyl ether
PubChem CID 51063134
IUPAC Name (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15549291
View Documents
MP Biomedicals Inc
0215549291
1 kg
Each for $863.93
Only null left
 
Description

Description

Specifications

Chemical Identifiers

9004-67-5
454.513
Cellulose methyl ether
(5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
C20H38O11
YLGXILFCIXHCMC-JHGZEJCSSA-N
51063134
COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifications

White
9004-67-5
Cellulose methyl ether
YLGXILFCIXHCMC-JHGZEJCSSA-N
(5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
51063134
Powder
C20H38O11
1 kg
COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
454.513
Methyl Cellulose, Viscosity of 2% aqueous solution at 25°C: 15cps
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at Room Temperature.

SDS
missing translation for 'documents'

missing translation for 'documents'