missing translation for 'onlineSavingsMsg'
Learn More

Methyl cellulose, viscosity 15 cPs

Catalog No. p-7059858
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4549022 100 g
AA4549036 500 g
AA45490A4 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4549022 Supplier Thermo Scientific Chemicals Supplier No. 04549022
Only null left

CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol

Methyl cellulose is used as an emulsifying and suspending agent in cosmetics, pharmaceutics and the chemical industry. It is used therapeutically as a bulk laxative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9004-67-5
Molecular Formula C20H38O11
Molecular Weight (g/mol) 454.513
MDL Number MFCD00081763
InChI Key YLGXILFCIXHCMC-JHGZEJCSSA-N
PubChem CID 51063134
IUPAC Name (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifications

Physical Form Powder
Merck Index 14,6040
Solubility Information Soluble in cold water.
Quantity 100 g
Odor Odorless
Chemical Name or Material Methyl cellulose, Viscosity: 15cPs
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only