missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Cellulose (350-550mPa.s, 2% in Water at 20°C), TCI America™
Supplier: TCI America M029325G
Specifications
| Methyl Cellulose (350-550mPa.s, 2% in Water at 20°C) | |
| White-Yellow | |
| MFCD00081763 | |
| YLGXILFCIXHCMC-JHGZEJCSSA-N | |
| (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane | |
| 51063134 |
| 9004-67-5 | |
| C20H38O11 | |
| 25 g | |
| COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC | |
| 454.513 | |
| Crystalline Powder |
Chemical Identifiers
| 9004-67-5 | |
| 454.513 | |
| YLGXILFCIXHCMC-JHGZEJCSSA-N | |
| (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane |
| C20H38O11 | |
| MFCD00081763 | |
| 51063134 | |
| COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC |
Safety and Handling
EINECSNumber : (8)-0186
RTECSNumber : FJ5959000
TSCA : Yes