Learn More
Methyl carbazate, 98%
CAS: 6294-89-9 | C2H6N2O2 | 90.08 g/mol
Supplier: Thermo Scientific Chemicals A145480E
Description
Methyl carbazate was used in the preparation of methyl 3-(4-methyl-benzyl-idene)carbazate. It was also used in the synthesis of imidazo[1,5-d][1,2,4]triazines. It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl carbazate was used in the preparation of methyl 3-(4-methyl-benzyl-idene)carbazate. It was also used in the synthesis of imidazo[1,5-d][1,2,4]triazines. It is used as pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| Methyl carbazate | |
| 70°C to 73°C | |
| C2H6N2O2 | |
| 2500 g | |
| 1679395 | |
| Soluble in water. | |
| COC(=O)NN | |
| 90.08 | |
| 90.08 |
| 6294-89-9 | |
| Weak | |
| MFCD00007594 | |
| UN2811 | |
| methyl carbazate, methyl hydrazinecarboxylate, methyl hydrazinocarboxylate, methoxycarbohydrazide, methylcarbazate, hydrazinecarboxylic acid, methyl ester, carbomethoxyhydrazide, methyl hydrazinoformate, methoxycarbonyl hydrazine, carbazic acid methyl ester | |
| WFJRIDQGVSJLLH-UHFFFAOYSA-N | |
| methyl N-aminocarbamate | |
| 80519 | |
| 98% |
Chemical Identifiers
| 6294-89-9 | |
| 90.08 | |
| WFJRIDQGVSJLLH-UHFFFAOYSA-N | |
| 80519 | |
| COC(=O)NN |
| C2H6N2O2 | |
| MFCD00007594 | |
| methyl carbazate, methyl hydrazinecarboxylate, methyl hydrazinocarboxylate, methoxycarbohydrazide, methylcarbazate, hydrazinecarboxylic acid, methyl ester, carbomethoxyhydrazide, methyl hydrazinoformate, methoxycarbonyl hydrazine, carbazic acid methyl ester | |
| methyl N-aminocarbamate |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P361-P363-P501c
H301+H311-H315-H319-H335-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 228-560-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only