missing translation for 'onlineSavingsMsg'
Learn More

Methyl Butyrate 99.0+%, TCI America™

Supplier:  TCI America B0763500ML

 View more versions of this product

Catalog No. B0763500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methyl Butyrate
Colorless
C5H10O2
500 mL
methyl butyrate, butanoic acid, methyl ester, methyl n-butyrate, butyric acid, methyl ester, methyl n-butanoate, butyric acid methyl ester, methyl-n-butyrate, n-butyric acid methyl ester, methyl butyrate natural, unii-cgx598508o
CCCC(=O)OC
102.13
102.13
Liquid
623-42-7
101°C
MFCD00009391
1237
UUIQMZJEGPQKFD-UHFFFAOYSA-N
methyl butanoate
12180
≥99.0% (GC)

Chemical Identifiers

623-42-7
102.13
UUIQMZJEGPQKFD-UHFFFAOYSA-N
12180
CCCC(=O)OC
C5H10O2
MFCD00009391
methyl butyrate, butanoic acid, methyl ester, methyl n-butyrate, butyric acid, methyl ester, methyl n-butanoate, butyric acid methyl ester, methyl-n-butyrate, n-butyric acid methyl ester, methyl butyrate natural, unii-cgx598508o
methyl butanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0779

RTECSNumber : ET5500000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions