missing translation for 'onlineSavingsMsg'
Learn More

Methyl Butyrate 99.0+%, TCI America™

Catalog No. B076325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B076325ML 25 mL
B0763500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B076325ML Supplier TCI America Supplier No. B076325ML

Chemical Identifiers

CAS 623-42-7
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00009391
InChI Key UUIQMZJEGPQKFD-UHFFFAOYSA-N
Synonym methyl butyrate, butanoic acid, methyl ester, methyl n-butyrate, butyric acid, methyl ester, methyl n-butanoate, butyric acid methyl ester, methyl-n-butyrate, n-butyric acid methyl ester, methyl butyrate natural, unii-cgx598508o
PubChem CID 12180
IUPAC Name methyl butanoate
SMILES CCCC(=O)OC

Specifications

Color Colorless
Boiling Point 101°C
Quantity 25 mL
UN Number 1237
Formula Weight 102.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Butyrate
EINECSNumber (2)-0779
RTECSNumber ET5500000
TSCA Yes