missing translation for 'onlineSavingsMsg'
Learn More

Methyl butyrate, 99%

Catalog No. AC414140050
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414140050 5 g Glass bottle
AC414141000 100 g Glass bottle
AC414142500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC414140050 Supplier Thermo Scientific Chemicals Supplier No. 414140050

CAS: 623-42-7 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 623-42-7
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00009391
InChI Key UUIQMZJEGPQKFD-UHFFFAOYSA-N
Synonym methyl butyrate, butanoic acid, methyl ester, methyl n-butyrate, butyric acid, methyl ester, methyl n-butanoate, butyric acid methyl ester, methyl-n-butyrate, n-butyric acid methyl ester, methyl butyrate natural, unii-cgx598508o
PubChem CID 12180
IUPAC Name methyl butanoate
SMILES CCCC(=O)OC

Specifications

Acidity 1mg KOH/g max.
Color Colorless to Yellow
Density 0.8970g/mL
Boiling Point 102°C
Flash Point 12°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3850 to 1.39
Linear Formula CH3(CH2)2COOCH3
Quantity 5 g
Specific Gravity 0.897
Merck Index 15, 6109
Solubility Information Solubility in water: insoluble
Formula Weight 102.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl butyrate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 210-792-1
RTECSNumber ET5500000
TSCA TSCA

RUO – Research Use Only