missing translation for 'onlineSavingsMsg'
Learn More

Methyl butyrate, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187593
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020190 1 mL
111018828 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020190 Supplier Merck Emd Millipore Supplier No. 193581ML
Only null left

Methyl butyrate is a clear colourless liquid which is slightly soluble in water. It adds flavour and odour to fruits and flowers. It has a characteristic pineapple odour.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 623-42-7
Vapor Pressure 40 mmHg (30°C)
Vapor Density 3.5 (vs. air)
Density 0.898 g/mL (at 25°C)
Flash Point 14°C
Physical Form Neat
Linear Formula CH3 CH2 CH2 COOCH3
MDL Number MFCD00009391
Boiling Point 102°C to 103°C
Type Analytical Standard
Quantity 1 mL
Formula Weight 102.13
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less