Learn More
Methyl butyrate, 98+%
CAS: 623-42-7 | C5H10O2 | 102.13 g/mol
Supplier: Thermo Scientific Chemicals L04250AP
Description
Methyl butyrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl butyrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
Specifications
| Methyl butyrate | |
| -85°C to -84°C | |
| 100°C to 102°C | |
| C5H10O2 | |
| MFCD00009391 | |
| UN1237 | |
| 14,6035 | |
| Slightly soluble in water. | |
| CCCC(=O)OC | |
| 102.13 | |
| 102.14 |
| 623-42-7 | |
| 0.898 | |
| 11°C (51°F) | |
| 1.386 | |
| 500 mL | |
| 1740743 | |
| methyl butyrate, butanoic acid, methyl ester, methyl n-butyrate, butyric acid, methyl ester, methyl n-butanoate, butyric acid methyl ester, methyl-n-butyrate, n-butyric acid methyl ester, methyl butyrate natural, unii-cgx598508o | |
| UUIQMZJEGPQKFD-UHFFFAOYSA-N | |
| methyl butanoate | |
| 12180 | |
| ≥98% |
Chemical Identifiers
| 623-42-7 | |
| 102.13 | |
| UUIQMZJEGPQKFD-UHFFFAOYSA-N | |
| 12180 | |
| CCCC(=O)OC |
| C5H10O2 | |
| MFCD00009391 | |
| methyl butyrate, butanoic acid, methyl ester, methyl n-butyrate, butyric acid, methyl ester, methyl n-butanoate, butyric acid methyl ester, methyl-n-butyrate, n-butyric acid methyl ester, methyl butyrate natural, unii-cgx598508o | |
| methyl butanoate |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL BUTYRATE
EINECSNumber : 210-792-1
RTECSNumber : ET5500000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only