missing translation for 'onlineSavingsMsg'
Learn More

Methyl bromoacetate, 99%

CAS: 96-32-2 | C3H5BrO2 | 152.98 g/mol

$73.48 - $795.43

Chemical Identifiers

CAS 96-32-2
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.98
MDL Number MFCD00000189
InChI Key YDCHPLOFQATIDS-UHFFFAOYSA-N
Synonym methyl bromoacetate, bromoacetic acid methyl ester, methyl monobromoacetate, acetic acid, bromo-, methyl ester, methyl alpha-bromoacetate, methylbromoacetate, acetic acid, 2-bromo-, methyl ester, methylester kyseliny bromoctove, methyl .alpha.-bromoacetate, methylester kyseliny bromoctove czech
PubChem CID 60984
IUPAC Name methyl 2-bromoacetate
SMILES COC(=O)CBr
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC169550250
SDS View Documents
Thermo Scientific Chemicals
169550250
25 g Glass bottle
Each for $73.48
Only null left
 
AC169551000
SDS View Documents
Thermo Scientific Chemicals
169551000
100 g Glass bottle
Each for $189.09
Only null left
 
AC169555000
SDS View Documents
Thermo Scientific Chemicals
169555000
500 g Glass bottle
Each for $795.43
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

96-32-2
152.98
YDCHPLOFQATIDS-UHFFFAOYSA-N
60984
COC(=O)CBr
C3H5BrO2
MFCD00000189
methyl bromoacetate, bromoacetic acid methyl ester, methyl monobromoacetate, acetic acid, bromo-, methyl ester, methyl alpha-bromoacetate, methylbromoacetate, acetic acid, 2-bromo-, methyl ester, methylester kyseliny bromoctove, methyl .alpha.-bromoacetate, methylester kyseliny bromoctove czech
methyl 2-bromoacetate

Specifications

96-32-2
1.6100g/mL
62°C
99%
C3H5BrO2
BrCH2CO2CH3
25 g
1.61
Solubility in water: practically insoluble. Other solubilities: soluble in methanol, ether and acetone
COC(=O)CBr
152.98
152.98
Liquid
Colorless to Yellow
145.0°C
Authentic
Glass bottle
1.4570 to 1.4590
MFCD00000189
02, 213
methyl bromoacetate, bromoacetic acid methyl ester, methyl monobromoacetate, acetic acid, bromo-, methyl ester, methyl alpha-bromoacetate, methylbromoacetate, acetic acid, 2-bromo-, methyl ester, methylester kyseliny bromoctove, methyl .alpha.-bromoacetate, methylester kyseliny bromoctove czech
YDCHPLOFQATIDS-UHFFFAOYSA-N
methyl 2-bromoacetate
60984
99%
Methyl bromoacetate, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 202-499-2

RTECSNumber : AF6300000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only