missing translation for 'onlineSavingsMsg'
Learn More

Methyl bromoacetate, 99%

CAS: 96-32-2 | C3H5BrO2 | 152.98 g/mol

Supplier:  Thermo Scientific Chemicals 169551000

 View more versions of this product

Catalog No. AC169551000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl bromoacetate
96-32-2
1.6100g/mL
62°C
99%
C3H5BrO2
BrCH2CO2CH3
100 g
1.61
Solubility in water: practically insoluble. Other solubilities: soluble in methanol, ether and acetone
COC(=O)CBr
152.98
152.98
Liquid
99%
Colorless to Yellow
145.0°C
Authentic
Glass bottle
1.4570 to 1.4590
MFCD00000189
02, 213
methyl bromoacetate, bromoacetic acid methyl ester, methyl monobromoacetate, acetic acid, bromo-, methyl ester, methyl alpha-bromoacetate, methylbromoacetate, acetic acid, 2-bromo-, methyl ester, methylester kyseliny bromoctove, methyl .alpha.-bromoacetate, methylester kyseliny bromoctove czech
YDCHPLOFQATIDS-UHFFFAOYSA-N
methyl 2-bromoacetate
60984
99%

Chemical Identifiers

96-32-2
152.98
YDCHPLOFQATIDS-UHFFFAOYSA-N
60984
COC(=O)CBr
C3H5BrO2
MFCD00000189
methyl bromoacetate, bromoacetic acid methyl ester, methyl monobromoacetate, acetic acid, bromo-, methyl ester, methyl alpha-bromoacetate, methylbromoacetate, acetic acid, 2-bromo-, methyl ester, methylester kyseliny bromoctove, methyl .alpha.-bromoacetate, methylester kyseliny bromoctove czech
methyl 2-bromoacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 202-499-2

RTECSNumber : AF6300000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only