missing translation for 'onlineSavingsMsg'
Learn More

Methyl-beta-D-xylopyranoside 96.0+%, TCI America™

Supplier:  TCI America M12535G

 View more versions of this product

Catalog No. M12535G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methyl-beta-D-xylopyranoside
155°C
C6H12O5
5 g
ZBDGHWFPLXXWRD-JGWLITMVSA-N
(2R,3R,4S,5R)-2-methoxyoxane-3,4,5-triol
11768891
164.16
Crystalline Powder
612-05-5
White-Yellow
MFCD00047532
methyl-beta-d-xylopyranoside, methyl beta-d-xyloside, methyl beta-d-xylopyranoside, beta-methyl-d-xyloside, 2r,3r,4s,5r-2-methoxytetrahydro-2h-pyran-3,4,5-triol, 1-o-methyl-beta-d-xylopyranose, 2r,3r,4s,5r-2-methoxyoxane-3,4,5-triol, 1-o-methyl-beta-d-xylopyranoside, beta-methylxyloside, beta-d-xylp-ome
COC1C(C(C(CO1)O)O)O
164.157
CHEBI:74863
≥96.0% (GC)

Chemical Identifiers

612-05-5
164.157
ZBDGHWFPLXXWRD-JGWLITMVSA-N
11768891
(2R,3R,4S,5R)-2-methoxyoxane-3,4,5-triol
C6H12O5
MFCD00047532
methyl-beta-d-xylopyranoside, methyl beta-d-xyloside, methyl beta-d-xylopyranoside, beta-methyl-d-xyloside, 2r,3r,4s,5r-2-methoxytetrahydro-2h-pyran-3,4,5-triol, 1-o-methyl-beta-d-xylopyranose, 2r,3r,4s,5r-2-methoxyoxane-3,4,5-triol, 1-o-methyl-beta-d-xylopyranoside, beta-methylxyloside, beta-d-xylp-ome
CHEBI:74863
COC1C(C(C(CO1)O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions