missing translation for 'onlineSavingsMsg'
Learn More

Methyl-beta-D-xylopyranoside 96.0+%, TCI America™

$132.90 - $394.92

Chemical Identifiers

CAS 612-05-5
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.157
MDL Number MFCD00047532
InChI Key ZBDGHWFPLXXWRD-JGWLITMVSA-N
Synonym methyl-beta-d-xylopyranoside, methyl beta-d-xyloside, methyl beta-d-xylopyranoside, beta-methyl-d-xyloside, 2r,3r,4s,5r-2-methoxytetrahydro-2h-pyran-3,4,5-triol, 1-o-methyl-beta-d-xylopyranose, 2r,3r,4s,5r-2-methoxyoxane-3,4,5-triol, 1-o-methyl-beta-d-xylopyranoside, beta-methylxyloside, beta-d-xylp-ome
PubChem CID 11768891
ChEBI CHEBI:74863
IUPAC Name (2R,3R,4S,5R)-2-methoxyoxane-3,4,5-triol
SMILES COC1C(C(C(CO1)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
M12535G
View Documents Promotion Details
N/A 5 g
Each for $132.90
Only null left
 
M125325G
View Documents Promotion Details
N/A 25 g
Each for $394.92
Only null left
 
Description

Description

Specifications

Chemical Identifiers

612-05-5
164.157
ZBDGHWFPLXXWRD-JGWLITMVSA-N
11768891
(2R,3R,4S,5R)-2-methoxyoxane-3,4,5-triol
C6H12O5
MFCD00047532
methyl-beta-d-xylopyranoside, methyl beta-d-xyloside, methyl beta-d-xylopyranoside, beta-methyl-d-xyloside, 2r,3r,4s,5r-2-methoxytetrahydro-2h-pyran-3,4,5-triol, 1-o-methyl-beta-d-xylopyranose, 2r,3r,4s,5r-2-methoxyoxane-3,4,5-triol, 1-o-methyl-beta-d-xylopyranoside, beta-methylxyloside, beta-d-xylp-ome
CHEBI:74863
COC1C(C(C(CO1)O)O)O

Specifications

612-05-5
White-Yellow
MFCD00047532
ZBDGHWFPLXXWRD-JGWLITMVSA-N
(2R,3R,4S,5R)-2-methoxyoxane-3,4,5-triol
11768891
164.16
Crystalline Powder
155°C
C6H12O5
methyl-beta-d-xylopyranoside, methyl beta-d-xyloside, methyl beta-d-xylopyranoside, beta-methyl-d-xyloside, 2r,3r,4s,5r-2-methoxytetrahydro-2h-pyran-3,4,5-triol, 1-o-methyl-beta-d-xylopyranose, 2r,3r,4s,5r-2-methoxyoxane-3,4,5-triol, 1-o-methyl-beta-d-xylopyranoside, beta-methylxyloside, beta-d-xylp-ome
COC1C(C(C(CO1)O)O)O
164.157
CHEBI:74863
≥96.0% (GC)
Methyl-beta-D-xylopyranoside
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents