missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl-beta-D-xylopyranoside 96.0+%, TCI America™
Supplier: TCI America M125325G
Specifications
| Methyl-beta-D-xylopyranoside | |
| 155°C | |
| C6H12O5 | |
| 25 g | |
| ZBDGHWFPLXXWRD-JGWLITMVSA-N | |
| (2R,3R,4S,5R)-2-methoxyoxane-3,4,5-triol | |
| 11768891 | |
| 164.16 | |
| Crystalline Powder |
| 612-05-5 | |
| White-Yellow | |
| MFCD00047532 | |
| methyl-beta-d-xylopyranoside, methyl beta-d-xyloside, methyl beta-d-xylopyranoside, beta-methyl-d-xyloside, 2r,3r,4s,5r-2-methoxytetrahydro-2h-pyran-3,4,5-triol, 1-o-methyl-beta-d-xylopyranose, 2r,3r,4s,5r-2-methoxyoxane-3,4,5-triol, 1-o-methyl-beta-d-xylopyranoside, beta-methylxyloside, beta-d-xylp-ome | |
| COC1C(C(C(CO1)O)O)O | |
| 164.157 | |
| CHEBI:74863 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 612-05-5 | |
| 164.157 | |
| ZBDGHWFPLXXWRD-JGWLITMVSA-N | |
| 11768891 | |
| (2R,3R,4S,5R)-2-methoxyoxane-3,4,5-triol |
| C6H12O5 | |
| MFCD00047532 | |
| methyl-beta-d-xylopyranoside, methyl beta-d-xyloside, methyl beta-d-xylopyranoside, beta-methyl-d-xyloside, 2r,3r,4s,5r-2-methoxytetrahydro-2h-pyran-3,4,5-triol, 1-o-methyl-beta-d-xylopyranose, 2r,3r,4s,5r-2-methoxyoxane-3,4,5-triol, 1-o-methyl-beta-d-xylopyranoside, beta-methylxyloside, beta-d-xylp-ome | |
| CHEBI:74863 | |
| COC1C(C(C(CO1)O)O)O |
Safety and Handling
TSCA : Yes