missing translation for 'onlineSavingsMsg'
Learn More

Methyl beta-D-xylopyranoside, 98%

CAS: 612-05-5 | C6H12O5 | 164.157 g/mol

Chemical Identifiers

CAS 612-05-5
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.157
MDL Number MFCD00047532
InChI Key ZBDGHWFPLXXWRD-JGWLITMVSA-N
Synonym methyl-beta-d-xylopyranoside, methyl beta-d-xyloside, methyl beta-d-xylopyranoside, beta-methyl-d-xyloside, 2r,3r,4s,5r-2-methoxytetrahydro-2h-pyran-3,4,5-triol, 1-o-methyl-beta-d-xylopyranose, 2r,3r,4s,5r-2-methoxyoxane-3,4,5-triol, 1-o-methyl-beta-d-xylopyranoside, beta-methylxyloside, beta-d-xylp-ome
PubChem CID 11768891
ChEBI CHEBI:74863
IUPAC Name (2R,3R,4S,5R)-2-methoxyoxane-3,4,5-triol
SMILES COC1C(C(C(CO1)O)O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ6672706
View Documents Promotion Details
N/A 5 g N/A
Only null left
 
AAJ6672709
View Documents Promotion Details
N/A 10 g N/A
Only null left
 
AAJ6672714
View Documents Promotion Details
N/A 25 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

612-05-5
164.157
ZBDGHWFPLXXWRD-JGWLITMVSA-N
11768891
(2R,3R,4S,5R)-2-methoxyoxane-3,4,5-triol
C6H12O5
MFCD00047532
methyl-beta-d-xylopyranoside, methyl beta-d-xyloside, methyl beta-d-xylopyranoside, beta-methyl-d-xyloside, 2r,3r,4s,5r-2-methoxytetrahydro-2h-pyran-3,4,5-triol, 1-o-methyl-beta-d-xylopyranose, 2r,3r,4s,5r-2-methoxyoxane-3,4,5-triol, 1-o-methyl-beta-d-xylopyranoside, beta-methylxyloside, beta-d-xylp-ome
CHEBI:74863
COC1C(C(C(CO1)O)O)O

Specifications

612-05-5
White
MFCD00047532
ZBDGHWFPLXXWRD-JGWLITMVSA-N
(2R,3R,4S,5R)-2-methoxyoxane-3,4,5-triol
11768891
164.16
Powder
155°C to 158°C
C6H12O5
methyl-beta-d-xylopyranoside, methyl beta-d-xyloside, methyl beta-d-xylopyranoside, beta-methyl-d-xyloside, 2r,3r,4s,5r-2-methoxytetrahydro-2h-pyran-3,4,5-triol, 1-o-methyl-beta-d-xylopyranose, 2r,3r,4s,5r-2-methoxyoxane-3,4,5-triol, 1-o-methyl-beta-d-xylopyranoside, beta-methylxyloside, beta-d-xylp-ome
COC1C(C(C(CO1)O)O)O
164.157
CHEBI:74863
98%
Methyl beta-D-xylopyranoside
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-289-7

TSCA : Yes

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only