missing translation for 'onlineSavingsMsg'
Learn More

Methyl beta-D-xylopyranoside, 98%

Catalog No. AAJ6672706
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6672706 5 g
AAJ6672709 10 g
AAJ6672714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6672706 Supplier Thermo Scientific Chemicals Supplier No. J6672706

CAS: 612-05-5 | C6H12O5 | 164.157 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 612-05-5
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.157
MDL Number MFCD00047532
InChI Key ZBDGHWFPLXXWRD-JGWLITMVSA-N
Synonym methyl-beta-d-xylopyranoside, methyl beta-d-xyloside, methyl beta-d-xylopyranoside, beta-methyl-d-xyloside, 2r,3r,4s,5r-2-methoxytetrahydro-2h-pyran-3,4,5-triol, 1-o-methyl-beta-d-xylopyranose, 2r,3r,4s,5r-2-methoxyoxane-3,4,5-triol, 1-o-methyl-beta-d-xylopyranoside, beta-methylxyloside, beta-d-xylp-ome
PubChem CID 11768891
ChEBI CHEBI:74863
IUPAC Name (2R,3R,4S,5R)-2-methoxyoxane-3,4,5-triol
SMILES COC1C(C(C(CO1)O)O)O

Specifications

Melting Point 155°C to 158°C
Color White
Quantity 5 g
Formula Weight 164.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Methyl beta-D-xylopyranoside
EINECSNumber 210-289-7
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only