missing translation for 'onlineSavingsMsg'
Learn More

Methyl Benzoylformate 97.0+%, TCI America™

Catalog No. p-7118816
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B103325G 25 g
B1033100G 100 g
B1033500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B103325G Supplier TCI America Supplier No. B103325G
Only null left

Chemical Identifiers

CAS 15206-55-0
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00008443
InChI Key YLHXLHGIAMFFBU-UHFFFAOYSA-N
Synonym methyl benzoylformate, methyl phenylglyoxylate, methyl oxophenylacetate, methyl phenylglyoxalate, benzoylformic acid methyl ester, methylbenzoylformate, phenylglyoxylic acid methyl ester, phenylglyoxylic acid, methyl ester, benzeneacetic acid, .alpha.-oxo-, methyl ester, unii-23f2045stg
PubChem CID 84835
ChEBI CHEBI:84256
IUPAC Name methyl 2-oxo-2-phenylacetate
SMILES COC(=O)C(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 251°C
Quantity 25 g
Formula Weight 164.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Benzoylformate
EINECSNumber (3)-3690
TSCA Yes